missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TC OT 39

Click to view available options
:
10mg
Description
TC OT 39 is a potent non-peptide oxytocin receptor partial agonist (EC50 values are 33 nM and 850 nM for the oxytocin receptor and V2 vasopressin receptor respectively). Also a V1a vasopressin receptor antagonist (Ki = 330 nM). Active in vivo in a rat model of uterine response.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | TC OT 39 |
| Purity | >97% |
| Molecular Weight (g/mol) | 600.78 |
| Inhibitors | TC OT 39 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction