missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TC-G 24

Product Code. 18055452
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18055452 50 mg 50mg
18054012 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18055452 Supplier Tocris Bioscience™ Supplier No. 4353/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective GSK-3β inhibitor

Potent GSK-3β inhibitor (IC50 = 17 nM); displays selectivity for GSK-3β over CDK2 (22% inhibition at 10 μM). Increases liver glycogen reserves in rodents. Brain penetrant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1257256-44-2
Molecular Formula C15H11ClN4O3

Specifications

Quantity 50 mg
Formula Weight 330.73
Purity >98%
Molecular Weight (g/mol) 330.728
Chemical Name or Material TC-G 24
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.