missing translation for 'onlineSavingsMsg'
Learn More

TC-G-1008, MedChemExpress

Product Code. 16490147
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490147 50 mg 50mg
16482846 25 mg 25mg
16473768 10 mg 10mg
16416497 5 mg 5mg
16404836 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16490147 Supplier Medchem Express Supplier No. HY103007
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress TC-G-1008 (GPR39-C3) is a potent and orally available GPR39 agonist with EC50 values of 0.4 and 0.8 nM for rat and human receptors respectively.

TC-G-1008 (GPR39-C3) is a potent and orally available GPR39 agonist with EC50 values of 0.4 and 0.8 nM for rat and human receptors respectively.

Specifications

Chemical Name or Material TC-G-1008
CAS 1621175-65-2
Color White
Purity Grade Notes Research
Molecular Formula C18H19ClN6O2S
Quantity 50 mg
Synonym GPR39-C3
Solubility Information DMSO : ≥ 100 mg/mL (238.72 mM)
SMILES CS(=O)(NC1=CC=C(CNC2=NC(NC)=NC(C3=NC=CC=C3)=C2)C(Cl)=C1)=O
Molecular Weight (g/mol) 418.9
Formula Weight 418.9
Percent Purity 99.03%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.