missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TC-G 1004
GREENER_CHOICE

Product Code. 30002000
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30002000 10 mg 10mg
30002014 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30002000 Supplier Tocris Bioscience™ Supplier No. 4407/10
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective A2A antagonist

Potent antagonist of adenosine A2A receptors; displays >100-fold selectivity for A2A over A1 receptors (Ki values are 0.44 and 85 nM respectively). Potentiates L-DOPA-induced rotational behavior in 6-OHDA-lesioned rats.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1061747-72-5
Molecular Formula C22H27N7O2

Specifications

Quantity 10 mg
Formula Weight 421.5
Purity >98%
Molecular Weight (g/mol) 421.505
Chemical Name or Material TC-G 1004
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.