missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TC-A 2317 hydrochloride

Description
Potent Aurora kinase A inhibitor (Ki = 1.2 nM compared to 101 nM for inhibition of Aurora kinase B). Selective over 60 other kinases (IC50 values > 1000 nM). Exhibits good cell permeability and antitumor activity.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 392.93 |
| Purity | >98% |
| Molecular Weight (g/mol) | 392.932 |
| Chemical Name or Material | TC-A 2317 hydrochloride |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?