missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ TC-100 Insect Medium, powder

Product Code. 10483432
Change view
Click to view available options
Quantity:
10 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10483432 10 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10483432 Supplier Gibco™ Supplier No. 43000033

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

TC-100 Insect Medium, powder

TC 100 Insect Medium was originally designed for the growth of insect cells, including Spodeptera frugiperda (Sf9), and the expression of recombinant Autographa Californica Nuclear Polyhedrosis Virus (rAcNPV) using the Baculovirus Expression Vector System (BEVS). We offer a variety of Gibco™ Media options for a range of insect cell culture applications. TC 100 Insect Medium is rich in amino acids and uses tryptose as an additional sugar source.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

cGMP Manufacturing and Quality System
Gibco™ TC 100 Insect Medium is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

Gibco™ TC 100 Insect Medium is commonly supplemented with 10% Fetal Bovine Serum (FBS) before use. TC 100 Insect Medium requires ambient CO2 in order to maintain physiological pH. The powder form of TC 100 Insect Medium requires the addition of 350 mg/L sodium bicarbonate before use.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Insect Cell
Form Powder
Product Type Insect Cell Medium, Powder
For Use With (Application) Insect cell culture
Quantity 10 L
Regulatory Status RUO
Content And Storage 2 to 8°C
Description TC-100 Insect Medium, 10mL, Powder, Insect Cells, Sf21, Sf9 Cell Line, 2 to 8°C Storage Temperature, Room Temperature, Ambient

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.