missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TC-1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-84292-0.1ML
This item is not returnable.
View return policy
Description
TC-1 Polyclonal specifically detects TC-1 in Human samples. It is validated for Western Blot.
Specifications
| TC-1 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 56892 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| chromosome 8 open reading frame 4, hTC-1, MGC22806, TC1 thyroid cancer-1, TC-1human thyroid cancer 1, Thyroid cancer protein 1 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Human TC-1 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction