missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBXA2R Polyclonal Antibody

TBXA2R Polyclonal Antibody
Brand: Invitrogen PA534259
This item is not returnable.
View return policy
Description
TBXA2R Polyclonal Antibody for IHC (P)
Specifications
| TBXA2R | |
| Polyclonal | |
| Unconjugated | |
| TBXA2R | |
| BDPLT13; prostanoid TP receptor; TBXA2R; Thromboxane A2 receptor; thromboxane prostanoid; Thromboxane receptor; TP; TXA2 receptor; TXA2-R; TXR2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6915 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P21731 | |
| TBXA2R | |
| Synthetic 17 amino acid peptide from C-terminus of human TBXA2R. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction