missing translation for 'onlineSavingsMsg'
Learn More

TBX5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£169.00 - £255.00

Specifications

Antigen TBX5
Dilution Immunohistochemistry-Paraffin 1:500-1:2000
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18637165
View Documents
Novus Biologicals
NBP2-99529-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18662236
View Documents
Novus Biologicals
NBP2-99529-50ul
50 μg
£169.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TBX5 Polyclonal antibody specifically detects TBX5 in Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

TBX5
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Mouse, Rat
HOS, T-box 5, T-box protein 5, T-box transcription factor TBX5
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human TBX5
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Immunohistochemistry-Paraffin 1:500-1:2000
Polyclonal
Purified
RUO
0.2 um filtered solution in PBS
6910
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.