missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBX22 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106540
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TBX22.
The T-box (TBX) motif is present in a family of genes whose structural features and expression patterns support their involvement in developmental gene regulation. The TBX gene family are largely conserved throughout metazoan evolution, and these genes code for putative transcription factors that share a uniquely defining DNA-binding domain. TBX genes are a family of developmental regulators with more than 20 members recently identified in invertebrates and vertebrates. Mutations in TBX genes are associated with the onset of several human diseases.
Specifications
| TBX22 | |
| Polyclonal | |
| Unconjugated | |
| TBX22 | |
| ABERS; CLPA; CPX; D230020M15Rik; dJ795G23.1; T-box 22; T-box protein 22; T-box transcription factor TBX22; TBOX22; Tbx 22; TBX22; TBXX | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 245572, 50945 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8K402, Q9Y458 | |
| TBX22 | |
| A synthesized peptide derived from human TBX22(Accession Q9Y458), corresponding to amino acid residues F10-K60. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction