missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBX22 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575266
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The T-box (TBX) motif is present in a family of genes whose structural features and expression patterns support their involvement in developmental gene regulation. The TBX gene family are largely conserved throughout metazoan evolution, and these genes code for putative transcription factors that share a uniquely defining DNA-binding domain. TBX genes are a family of developmental regulators with more than 20 members recently identified in invertebrates and vertebrates. Mutations in TBX genes are associated with the onset of several human diseases.
Specifications
| TBX22 | |
| Polyclonal | |
| Unconjugated | |
| TBX22 | |
| ABERS; CLPA; CPX; D230020M15Rik; dJ795G23.1; T-box 22; T-box protein 22; T-box transcription factor TBX22; TBOX22; Tbx 22; TBX22; TBXX | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 245572, 302369, 50945 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8K402, Q9Y458 | |
| TBX22 | |
| Synthetic peptide corresponding to the N-terminual of Human TBX22. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction