missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBX22 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-86851
This item is not returnable.
View return policy
Description
TBX22 Polyclonal specifically detects TBX22 in Mouse samples. It is validated for Western Blot.
Specifications
| TBX22 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| cleft palate and/or ankyloglossia, CLPA, CPX, dJ795G23.1, T-box 22, T-box protein 22, T-box transcription factor TBX22, TBOX22, TBXX | |
| The immunogen is a synthetic peptide corresponding to a region of Mouse TBX22 | |
| 100 μL | |
| Stem Cell Markers | |
| 50945 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction