missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBX22 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94171-0.02ml
This item is not returnable.
View return policy
Description
TBX22 Polyclonal antibody specifically detects TBX22 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| TBX22 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| cleft palate and/or ankyloglossia, CLPA, CPX, dJ795G23.1, T-box 22, T-box protein 22, T-box transcription factor TBX22, TBOX22, TBXX | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 291-520 of human TBX22 (NP_058650.1). LDGLLETYPWRPSFTLDFKTFGADTQSGSSGSSPVTSSGGAPSPLNSLLSPLCFSPMFHLPTSSLGMPCPEAYLPNVNLPLCYKICPTNFWQQQPLVLPAPERLASSNSSQSLAPLMMEVPMLSSLGVTNSKSGSSEDSSDQYLQAPNSTNQMLYGLQSPGNIFLPNSITPEALSCSFHPSYDFYRYNFSMPSRLISGSNHLKVNDDSQVSFGEGKCNHVHWYPAINHYL | |
| 0.02 mL | |
| Stem Cell Markers | |
| 50945 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction