missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBX20 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107034
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
TBX20 is a member of the T-box transcription factor family expressed in the developing heart, eye, ventral neural tube, and limbs, indicating a possible role in regulating development of these tissues.
Specifications
| TBX20 | |
| Polyclonal | |
| Unconjugated | |
| TBX20 | |
| 9430010M06Rik; AL022859; ASD4; cb826; H15-related T-box transcription factor hrT; hrt; T-box 20; T-box protein 20; T-box transcription factor; T-box transcription factor 20; T-box transcription factor TBX20; T-box transcription factor TBX20-like; Tbx12; TBX20; whz; ZF-TBX20 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57057 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q9UMR3 | |
| TBX20 | |
| Synthetic peptide directed towards the middle region of human TBX20 (aa 200-250). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction