Learn More
Invitrogen™ TBX20 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5113564
Product Code. 17141424
  • £437.00 / 100µL
Description
Immunogen sequence: RANAFSIAAL MSSGGSKEKE ATENTIKPLE QFVEKSSCAQ PLGELTSLDA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.
TBX20 is a member of the T-box transcription factor family expressed in the developing heart, eye, ventral neural tube, and limbs, indicating a possible role in regulating development of these tissues.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TBX20
Polyclonal
Unconjugated
TBX20
9430010M06Rik; AL022859; ASD4; cb826; H15-related T-box transcription factor hrT; hrt; T-box 20; T-box protein 20; T-box transcription factor; T-box transcription factor 20; T-box transcription factor TBX20; T-box transcription factor TBX20-like; Tbx12; TBX20; whz; ZF-TBX20
Rabbit
Affinity Chromatography
RUO
57057
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9UMR3
TBX20
Synthetic peptide directed towards the N-terminal region of Human TBX20 (aa 14-63).