missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBX20 Antibody (OTI5D4), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
£195.00 - £417.00
Specifications
| Antigen | TBX20 |
|---|---|
| Clone | OTI5D4 |
| Dilution | Western Blot 1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306286
|
Novus Biologicals
NBP3-44934-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306528
|
Novus Biologicals
NBP3-44934-30ul |
30 μL |
£195.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TBX20 Monoclonal antibody specifically detects TBX20 in Human,Mouse samples. It is validated for Western BlotSpecifications
| TBX20 | |
| Western Blot 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ASD4, T-box 20, T-box protein 20, T-box transcription factor TBX20 | |
| Human recombinant protein fragment corresponding to amino acids 1-340 of human TBX20(NP_001071121) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| OTI5D4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cardiovascular Biology | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 57057 | |
| IgG1 | |
| >95% | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title