missing translation for 'onlineSavingsMsg'
Learn More

TBX20 Antibody (OTI2C7) [mFluor Violet 450 SE], Novus Biologicals™

Product Code. 30687354 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30687354 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30687354 Supplier Novus Biologicals Supplier No. NBP344736MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TBX20 Monoclonal antibody specifically detects TBX20 in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBX20
Applications Western Blot
Classification Monoclonal
Clone OTI2C7
Conjugate mFluor Violet 450 SE
Formulation 50mM Sodium Borate
Gene Alias ASD4, T-box 20, T-box protein 20, T-box transcription factor TBX20
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-340 of human TBX20(NP_001071121) produced in E.coli.
Purification Method >95%
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 57057
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.