missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBX2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22346-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene product is the human homolog of mouse Tbx2, and shares strong sequence similarity with Drosophila omb protein. Expression studies indicate that this gene may have a potential role in tumorigenesis as an immortalizing agent. Transcript heterogeneity due to alternative polyadenylation has been noted for this gene.Specifications
| TBX2 | |
| Polyclonal | |
| Unconjugated | |
| TBX2 | |
| T box 2, T box protein 2, TBX2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21385, 6909 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13207, Q60707 | |
| TBX2 | |
| TBX2 Fusion Protein Ag17860 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction