missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBX2 Monoclonal Antibody (393725)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561512
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene product is the human homolog of mouse Tbx2, and shares strong sequence similarity with Drosophila omb protein. Expression studies indicate that this gene may have a potential role in tumorigenesis as an immortalizing agent. Transcript heterogeneity due to alternative polyadenylation has been noted for this gene.
Specifications
| TBX2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q13207 | |
| TBX2 | |
| E. coli-derived recombinant human TBX2, Thr593-Arg702. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 393725 | |
| Unconjugated | |
| TBX2 | |
| FLJ10169; LOW QUALITY PROTEIN: T-box transcription factor TBX2; T-box 2; T-box 2 protein; T-box 2 transcription factor; T-box protein 2; T-box transcription factor 2; T-box transcription factor TBX2; TBX2 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 6909 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction