missing translation for 'onlineSavingsMsg'
Learn More

TBX18 Antibody (4D3), Novus Biologicals™

Product Code. 18379238 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379238 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379238 Supplier Novus Biologicals Supplier No. H00009096M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TBX18 Monoclonal antibody specifically detects TBX18 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBX18
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 4D3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100 to 1:2000, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. XP_496819
Gene Alias T-box 18, T-box protein 18, T-box transcription factor TBX18
Host Species Mouse
Immunogen TBX18 (XP_496819, 454 a.a. ~ 560 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. STLLQGTGNGVPATHPHLLSGSSCSSPAFHLGPNTSQLCSLAPADYSACARSGLTLNRYSTSLAETYNRLTNQAGETFAPPRTPSYVGVSSSTSVNMSMGGTDGDTF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9096
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.