missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBX15/TBX18 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538563
This item is not returnable.
View return policy
Description
T-box transcription factors are a group of phylogenetically conserved genes that contain a uniquely defining DNA-binding domain, the T-box domain. These genes are believed to be involved in the regulation of development processes. In mouse embryo development, Tbx15 is expressed at the beginning of day 9.5, primarily in the craniofacial region and in the developing limbs. Twelve human T-box genes have been isolated so far. TBX 18 has been reported to be involved in numerous development processes and to act as an antiapoptotic factor.
Specifications
| TBX15/TBX18 | |
| Polyclonal | |
| Unconjugated | |
| TBX15 | |
| T-box transcription factor TBX18, T-box protein 18, T-box transcription factor TBX15, T-box protein 15 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 21384, 6913, 76365, 9096 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95935, Q96SF7, O70306, Q9EPZ6 | |
| TBX15, TBX18 | |
| A synthetic peptide derived from the internal region of human TBX15/18 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction