missing translation for 'onlineSavingsMsg'
Learn More

TBX15 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£222.00 - £484.00

Specifications

Antigen TBX15
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18607535
View Documents
Novus Biologicals
NBP2-49036-25ul
25 μL
£222.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18663169
View Documents
Novus Biologicals
NBP2-49036
0.1 mL
£484.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TBX15 Polyclonal antibody specifically detects TBX15 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

TBX15
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
T-box 14, T-box 15, T-box protein 14, T-box protein 15, T-box transcription factor TBX14, T-box transcription factor TBX15, TBX14
This antibody was developed against a recombinant protein corresponding to amino acids: CSPPTFHLAPNTFNVGCRESQLCNLNLSDYPPCARSNMAALQSYPGLSDSGYNRLQSGTTSATQPSETFMPQRTPSLISGIPTPPSLPGNSKMEAYGGQLGSFPTSQFQYVMQ
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Purified
RUO
PBS (pH 7.2), 40% Glycerol
6913
IgG
Immunogen affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.