missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBX10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561615
This item is not returnable.
View return policy
Description
Immunogen sequence: PARSHSSLSP CVLKGATDRE KDPNKASAST SKTPAWLHHQ LLPPPEVLLA PATYRPVTYQ SLYSGAPSHL GIPRTR Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 64%.
The T-box (Tbx) motif is present in a family of genes whose structural features and expression patterns support their involvement in developmental gene regulation. The Tbx gene family are largely conserved throughout metazoan evolution, and these genes code for putative transcription factors that share a uniquely defining DNA-binding domain. Tbx genes are a family of developmental regulators with more than 20 members recently identified in invertebrates and vertebrates. Mutations in Tbx genes are associated with the onset of several human diseases. Our understanding of functional mechanisms of Tbx products has come mainly from the prototypical T/Brachyury, which is a transcription activator.
Specifications
| TBX10 | |
| Polyclonal | |
| Unconjugated | |
| TBX10 | |
| dancer; Dc; mmTBX7; T-box 10; T-box 10, pseudogene 1; T-box 13; T-box 7; T-box protein 10; T-box protein 13; T-box transcription factor TBX10; T-box-containing transcriptional activator; TBX10; Tbx10-ps1; Tbx13; TBX7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 347853 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O75333 | |
| TBX10 | |
| Recombinant protein corresponding to Human TBX10. Recombinant protein control fragment (Product #RP-99864). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto