missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBX10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538541
This item is not returnable.
View return policy
Description
The T-box (Tbx) motif is present in a family of genes whose structural features and expression patterns support their involvement in developmental gene regulation. The Tbx gene family are largely conserved throughout metazoan evolution, and these genes code for putative transcription factors that share a uniquely defining DNA-binding domain. Tbx genes are a family of developmental regulators with more than 20 members recently identified in invertebrates and vertebrates. Mutations in Tbx genes are associated with the onset of several human diseases. Our understanding of functional mechanisms of Tbx products has come mainly from the prototypical T/Brachyury, which is a transcription activator.
Specifications
| TBX10 | |
| Polyclonal | |
| Unconjugated | |
| TBX10 | |
| T-box transcription factor TBX10, T-box protein 10, TBX10, TBX7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 347853 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75333 | |
| TBX10 | |
| A synthetic peptide derived from the internal region of human TBX10 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction