missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBX1 Antibody (OTI1C2), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74470CL1
This item is not returnable.
View return policy
Description
TBX1 Monoclonal antibody specifically detects TBX1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| TBX1 | |
| Monoclonal | |
| CoraFluor 1 | |
| brachyury, CAFS, CTHM, DGCR, DGS, DORV, T-box 1, T-box 1 transcription factor C, T-box protein 1, T-box transcription factor TBX1, TBX1C, Testis-specific T-box protein, TGA, VCFS | |
| Full length human recombinant protein of human TBX1(NP_542377) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot | |
| OTI1C2 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 6899 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction