missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ 20X TBS Wash Buffer for IHC/ICC

Product Code. 15556276
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15556276 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15556276 Supplier Invitrogen™ Supplier No. 00495456
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in immunohistochemistry and immunocytochemistry protocols which require washing to remove non-specific binding. TBS Wash Buffer for IHC/ICC is recommended for eFluor™ Nanocrystal conjugated-antibodies following antibody incubation.

  • Compatible with organic dye-conjugated antibodies as well as unconjugated antibodies that may be used in two and three step staining protocols
  • The buffer is supplied as a 20x stock solution that should be diluted to 1x with distilled water
TRUSTED_SUSTAINABILITY

Specifications

Product Type Wash Buffer
Quantity 100 mL
For Use With (Application) Immunocytochemistry (ICC), Immunohistochemistry (IHC)
Product Line eBioscience
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.