missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBRG4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10177-2-AP-150UL
This item is not returnable.
View return policy
Description
TBRG4 (transforming growth factor beta regulator 4), also known as CPR2 (cell cycle progression restoration protein 2) or FAST 4 (FAST kinase domain-containing protein 4), is a 631 amino acid protein that contains one RAP domain and belongs to the FAST kinase family. TBRG4 is ubiquitously expressed and may have a role in cell cycle progression. Existing as two alternatively spliced isoforms, the gene encoding TBRG4 maps to human chromosome 7p13.Specifications
| TBRG4 | |
| Polyclonal | |
| Unconjugated | |
| TBRG4 | |
| CPR2, FASTKD4, KIAA0948, Protein TBRG4, TBRG4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9238 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.62 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q969Z0 | |
| TBRG4 | |
| TBRG4 Fusion Protein Ag0212 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction