missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBRG4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584006
This item is not returnable.
View return policy
Description
Immunogen sequence: FHQLLCLLNS QIASVWHGTL SKLLGSLYAL GIPKASKELQ SVEQEVRWRM RKLKYKHLAF LAESCATLSQ EQHSQELLAE LLTHLERRWT EIEDSHTLVT VMMKVG Highest antigen sequence indentity to the following orthologs: Rat - 72%, Mouse - 73%.
TBRG4 (transforming growth factor beta regulator 4), also known as CPR2 (cell cycle progression restoration protein 2) or FAST 4 (FAST kinase domain-containing protein 4), is a 631 amino acid protein that contains one RAP domain and belongs to the FAST kinase family. TBRG4 is ubiquitously expressed and may have a role in cell cycle progression. Existing as two alternatively spliced isoforms, the gene encoding TBRG4 maps to human chromosome 7p13.
Specifications
| TBRG4 | |
| Polyclonal | |
| Unconjugated | |
| TBRG4 | |
| 2310042P22Rik; AA120735; AA408001; AI527316; cell cycle progression 2 protein; cell cycle progression protein 2; Cell cycle progression restoration protein 2; CPR2; FAST 4; FAST kinase domain-containing protein 4; FAST kinase domains 4; FASTKD4; H_TD2522F11.8; KIAA0948; mKIAA0948; Protein TBRG4; R74877; Tb-12; TBRG4; transforming growth factor beta regulated gene 4; transforming growth factor beta regulator 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9238 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q969Z0 | |
| TBRG4 | |
| Recombinant protein corresponding to Human TBRG4. Recombinant protein control fragment (Product #RP-104899). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction