missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TBRG4 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA554236

Product Code. 15732171

  • £446.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: HLVKRCTCLL REAARQAPAM APVGRLRLAW VAHKTLTSSA TSPISHLPGS LMEPVEKERA STPYIEKQVD HLIKKATRPE ELLELLGGSH DLDSNQAAMV LIRLSHLLSE KP Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 68%.

TBRG4 (transforming growth factor beta regulator 4), also known as CPR2 (cell cycle progression restoration protein 2) or FAST 4 (FAST kinase domain-containing protein 4), is a 631 amino acid protein that contains one RAP domain and belongs to the FAST kinase family. TBRG4 is ubiquitously expressed and may have a role in cell cycle progression. Existing as two alternatively spliced isoforms, the gene encoding TBRG4 maps to human chromosome 7p13.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TBRG4
Polyclonal
Unconjugated
TBRG4
2310042P22Rik; AA120735; AA408001; AI527316; cell cycle progression 2 protein; cell cycle progression protein 2; Cell cycle progression restoration protein 2; CPR2; FAST 4; FAST kinase domain-containing protein 4; FAST kinase domains 4; FASTKD4; H_TD2522F11.8; KIAA0948; mKIAA0948; Protein TBRG4; R74877; Tb-12; TBRG4; transforming growth factor beta regulated gene 4; transforming growth factor beta regulator 4
Rabbit
Antigen Affinity Chromatography
RUO
9238
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.4 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q969Z0
TBRG4
Recombinant protein corresponding to Human TBRG4. Recombinant protein control fragment (Product #RP-92047).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.