missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBRG1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13346-1-AP-150UL
This item is not returnable.
View return policy
Description
TBRG1 acts as a growth inhibitor. It can activate TP53/p53, cause G1 arrest and collaborate with CDKN2A to restrict proliferation, but it does not require either protein to inhibit DNA synthesis. TBRG1 redistributes CDKN2A into the nucleoplasm, and is involved in maintaining chromosomal stability.Specifications
| TBRG1 | |
| Polyclonal | |
| Unconjugated | |
| TBRG1 | |
| NIAM, TB 5, TBRG1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84897 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3YBR2 | |
| TBRG1 | |
| TBRG1 Fusion Protein Ag4179 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction