missing translation for 'onlineSavingsMsg'
Learn More

TBRG1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18352176 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18352176 20 μL 20µL
18330457 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18352176 Supplier Novus Biologicals Supplier No. NBP30486420UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TBRG1 Polyclonal specifically detects TBRG1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBRG1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias FLJ14621, FLJ90113, MGC129890, NIAMFLJ25020, Nuclear interactor of ARF and Mdm2, TB-5, transforming growth factor beta regulator 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 22-150 of human TBRG1 (NP_116200.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84897
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.