missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20932-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene is a member of a conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similar protein has been disrupted in mice and shown to be critical for early cortical development, and causes loss of projection neurons in the olfactory bulbs and olfactory cortex. The C-terminal region this similar protein was found to be necessary and sufficient for association with the guanylate kinase domain of calcium/calmodulin-dependent serine protein kinase.Specifications
| TBR1 | |
| Polyclonal | |
| Unconjugated | |
| TBR1 | |
| T box brain protein 1, T box, brain, 1, T brain 1, TBR 1, TBR1, TES 56 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10716, 21375, 680427 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16650, Q64336 | |
| Tbr1 | |
| TBR1 Fusion Protein Ag14935 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction