Learn More
TBR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20932-1-AP-150UL
 View more versions of this product
Product Code. 16860515
  • £349.00 / 150µL
Description
This gene is a member of a conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similar protein has been disrupted in mice and shown to be critical for early cortical development, and causes loss of projection neurons in the olfactory bulbs and olfactory cortex. The C-terminal region this similar protein was found to be necessary and sufficient for association with the guanylate kinase domain of calcium/calmodulin-dependent serine protein kinase.
Specifications
Specifications
TBR1
Polyclonal
Unconjugated
TBR1
T box brain protein 1, T box, brain, 1, T brain 1, TBR 1, TBR1, TES 56
Rabbit
Antigen Affinity Chromatography
RUO
10716, 21375, 680427
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q16650, Q64336
Tbr1
TBR1 Fusion Protein Ag14935
150 μL
Primary
Human, Mouse, Rat