missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114784
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TBR1.
This gene is a member of a conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similar protein has been disrupted in mice and shown to be critical for early cortical development, and causes loss of projection neurons in the olfactory bulbs and olfactory cortex. The C-terminal region this similar protein was found to be necessary and sufficient for association with the guanylate kinase domain of calcium/calmodulin-dependent serine protein kinase.
Specifications
| TBR1 | |
| Polyclonal | |
| Unconjugated | |
| TBR1 | |
| LOW QUALITY PROTEIN: T-box brain protein 1; T-box brain gene 1; T-box brain protein 1; T-box brain transcription factor 1; T-box, brain 1; T-box, brain, 1; T-box, brain, 1-like; TBR1; TBR-1; T-brain-1; TES-56 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10716, 21375, 680427 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q16650, Q64336 | |
| TBR1 | |
| A synthesized peptide derived from human TBR1(Accession Q16650), corresponding to amino acid residues P38-D88. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction