missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585065
This item is not returnable.
View return policy
Description
Immunogen sequence: LEHCLSPSIM LSKKFLNVSS SYPHSGGSEL VLHDHPIIST TDNLERSSPL KKITRGMTNQ SDTDNFPDSK DSPGDVQRSK LSPVLDGVSE LRHSFD Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
This gene is a member of a conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. A similar protein has been disrupted in mice and shown to be critical for early cortical development, and causes loss of projection neurons in the olfactory bulbs and olfactory cortex. The C-terminal region this similar protein was found to be necessary and sufficient for association with the guanylate kinase domain of calcium/calmodulin-dependent serine protein kinase.
Specifications
| TBR1 | |
| Polyclonal | |
| Unconjugated | |
| TBR1 | |
| LOW QUALITY PROTEIN: T-box brain protein 1; T-box brain gene 1; T-box brain protein 1; T-box brain transcription factor 1; T-box, brain 1; T-box, brain, 1; T-box, brain, 1-like; TBR1; TBR-1; T-brain-1; TES-56 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10716 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16650 | |
| TBR1 | |
| Recombinant protein corresponding to Human TBR1. Recombinant protein control fragment (Product #RP-106332). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction