Learn More
Invitrogen™ TBPL1 Recombinant Rabbit Monoclonal Antibody (24GB4060)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557282
 View more versions of this product
Product Code. 30274959
  • £126.00 / 20µL
Description
TBPL1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes a protein that serves the same function as TBP and substitutes for TBP at some promoters that are not recognized by TFIID. It is essential for spermiogenesis and believed to be important in expression of developmentally regulated genes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TBPL1
Recombinant Monoclonal
Unconjugated
TBPL1
21 kDa TBP-like protein; 21-kDA TBP-like protein; 4732475G08; AW011832; AW491032; D18347; MGC:8389; MGC:9620; second TBP of unique DNA protein; STD; STUD; TATA box binding protein-like 1; TATA box binding protein-like protein; TATA box-binding protein related factor 2; TATA box-binding protein-like 1; TATA box-binding protein-like protein 1; TATA box-binding protein-like protein 1; TATA box-binding protein-like protein 1-like protein; TATA box-binding protein-related factor 2; TATA-box binding protein like 1; tbpl1; TBP-like 1; TBP-like factor; TBP-like protein; TBP-like protein 1; TBP-related factor 2; TBP-related protein; Tlf; TLP; Tlp21; trf; trf2; TRP; Unknown (protein for MGC:134257); zgc:92340
Rabbit
Affinity chromatography
RUO
689030, 9519
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4060
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P62380
TBPL1
A synthesized peptide derived from human TBPL1 (aa 100-150).
20 μL
Primary
Human, Rat