missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBPL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12258-1-AP-20UL
This item is not returnable.
View return policy
Description
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes a protein that serves the same function as TBP and substitutes for TBP at some promoters that are not recognized by TFIID. It is essential for spermiogenesis and believed to be important in expression of developmentally regulated genes.Specifications
| TBPL1 | |
| Polyclonal | |
| Unconjugated | |
| TBPL1 | |
| 21 kDa TBP like protein, STUD, TBP like 1, TBP like factor, TBP like protein 1, TBP related factor 2, TBP related protein, TBPL1, TLF, TLP, TLP21, TRF2, TRP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 237336, 689030, 9519 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62340, P62380 | |
| TBPL1 | |
| TBPL1 Fusion Protein Ag2880 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction