missing translation for 'onlineSavingsMsg'
Learn More

TBPE (Tetrabromophenolphthalein Ethyl Ester Potassium Salt), TRC

Product Code. 30427191
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
30427191 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 30427191

Brand: Toronto Research Chemicals TRCT104030100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material TBPE (Tetrabromophenolphthalein Ethyl Ester Potassium Salt)
CAS 62637-91-6
Molecular Formula C22 H13 Br4 O4 . K
Quantity 100 mg
Synonym Benzoic acid','2-[(3,5-dibromo-4-hydroxyphenyl)(3,5-dibromo-4-oxo-2,5-cyclohexadien-1-ylidene)methyl]-','ethyl ester','potassium salt (9CI), Phenolphthalein','3',3'',5',5''-tetrabromo-','ethyl ester','potassium deriv. (6CI)
InChI Formula InChI=1S/C22H14Br4O4.K/c1-2-30-22(29)14-6-4-3-5-13(14)19(11-7-15(23)20(27)16(24)8-11)12-9-17(25)21(28)18(26)10-12;/h3-10,27H,2H2,1H3;/q;+1/p-1
SMILES [K+].CCOC(=O)c1ccccc1C(=C2C=C(Br)C(=O)C(=C2)Br)c3cc(Br)c([O-])c(Br)c3
IUPAC Name potassium;2,6-dibromo-4-[(3,5-dibromo-4-oxocyclohexa-2,5-dien-1-ylidene)-(2-ethoxycarbonylphenyl)methyl]phenolate
Molecular Weight (g/mol) 700.051
Formula Weight 695.718
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.