missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tbp7 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00 - £366.00
Specifications
| Antigen | Tbp7 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18627491
|
Novus Biologicals
NBP2-93919-0.02ml |
0.02 mL |
£149.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681441
|
Novus Biologicals
NBP2-93919-0.1ml |
0.1 mL |
£366.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tbp7 Polyclonal antibody specifically detects Tbp7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Tbp7 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| PBS (pH 7.3), 50% glycerol | |
| 5704 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| MB67 interacting protein, MGC13687,26S protease regulatory subunit 6B, MGC23214, MGC8570, protease 26S subunit 6, proteasome (prosome, macropain) 26S subunit, ATPase, 4,26S proteasome AAA-ATPase subunit RPT3, Proteasome 26S subunit ATPase 4, S6Tat-binding protein 7, TBP-7MB67-interacting protein, TBP7MIP224 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-165 of human PSMC4 (NP_006494.1). MEEIGILVEKAQDEIPALSVSRPQTGLSFLGPEPEDLEDLYSRYKKLQQELEFLEVQEEYIKDEQKNLKKEFLHAQEEVKRIQSIPLVIGQFLEAVDQNTAIVGSTTGSNYYVRILSTIDRELLKPNASVALHKHSNALVDVLPPEADSSIMMLTSDQKPDVMYA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title