missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tbp7 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99612AF350
This item is not returnable.
View return policy
Description
Tbp7 Polyclonal antibody specifically detects Tbp7 in Human samples. It is validated for Western Blot
Specifications
| Tbp7 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 350 | |
| MB67 interacting protein, MGC13687,26S protease regulatory subunit 6B, MGC23214, MGC8570, protease 26S subunit 6, proteasome (prosome, macropain) 26S subunit, ATPase, 4,26S proteasome AAA-ATPase subunit RPT3, Proteasome 26S subunit ATPase 4, S6Tat-binding protein 7, TBP-7MB67-interacting protein, TBP7MIP224 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Tbp7. | |
| 0.1 mL | |
| Neuroscience, Ubiquitin Proteasome Pathway | |
| 5704 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction