missing translation for 'onlineSavingsMsg'
Learn More

TBP Mouse anti-Human, Mouse, Porcine, Rat, Clone: 2H3B2, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66166-1-IG-150UL

Product Code. 16837795

  • £189.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody detects human TBP (∽40 kDa) and mouse/rat Tbp (∽35 kDa).

RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA, RAC1, and CDC42.
Specifications

Specifications

TBP
Monoclonal
1.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P20226, P29037
TBP
TBP Fusion Protein Ag12383
150 μL
Primary
Human, Mouse, Pig, Rat
Antibody
IgG2a
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
2H3B2
Unconjugated
TBP
GTF2D, GTF2D1, SCA17, TATA binding factor, TATA box binding protein, TATA box factor, TATA sequence binding protein, TBP, TF2D, TFIID
Mouse
Protein A
RUO
110259740, 117526, 21374, 6908
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.