missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TBP Monoclonal Antibody (1TBP18)
GREENER_CHOICE

Product Code. 11593463
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11593463 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11593463 Supplier Invitrogen™ Supplier No. MA110883

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10883 detects TATA-Binding Protein from human, mouse, and rat samples. It does not react with TBP from Drosophila, yeast, silkworms or xenopus. MA1-10883 has been successfully used in Western blot and immunoprecipitation applications. The MA1-10883 immunogen is N-terminal domain of the human TATA-binding protein (TBP).

RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA, RAC1, and CDC42.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBP
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1TBP18
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Tbp
Gene Accession No. P20226, P29037
Gene Alias Gtf2d; GTF2D1; HDL4; MGC117320; MGC126054; MGC126055; RP1-191N21.3; SCA17; TATA binding protein; TA-TA binding protein 1; TATA box binding protein; TATA sequence-binding protein; TATA-binding factor; TATA-binding protein; TATA-box binding protein; TATA-box binding protein N-terminal domain; TATA-box factor; tata-box-binding; TATA-box-binding protein; TBP; TBP0; TBP1; TF2D; TFIID; TFIID core protein; transcription initiation factor TFIID TBP subunit; Unknown (protein for MGC:138100)
Gene Symbols Tbp
Host Species Mouse
Immunogen N-terminal domain of the human TATA-binding protein (TBP).
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117526, 21374, 6908
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.