missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBP like protein TLP Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93267-0.02ml
This item is not returnable.
View return policy
Description
TBP like protein TLP Polyclonal antibody specifically detects TBP like protein TLP in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| TBP like protein TLP | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| 21 kDa TBP-like protein, STUD21-kDA TBP-like protein, TATA box binding protein-related factor 2, TATA box-binding protein-like protein 1, TATA box-binding protein-related factor 2, TBP-like 1, TBP-like factor, TBP-like protein 1, TBP-related factor 2, TBP-related protein, TLFMGC:8389, TLP21, TLPMGC:9620, TRF2Second TBP of unique DNA protein, TRP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-186 of human TBPL1 (NP_004856.1). MDADSDVALDILITNVVCVFRTRCHLNLRKIALEGANVIYKRDVGKVLMKLRKPRITATIWSSGKIICTGATSEEEAKFGARRLARSLQKLGFQVIFTDFKVVNVLAVCNMPFEIRLPEFTKNNRPHASYEPELHPAVCYRIKSLRATLQIFSTGSITVTGPNVKAVATAVEQIYPFVFESRKEIL | |
| 0.02 mL | |
| Neurodegeneration, Neuroscience | |
| 9519 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction