missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBLR1 Recombinant Rabbit Monoclonal Antibody (23GB4935), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552746
This item is not returnable.
View return policy
Description
Chromatin organization plays a fundamental role in regulating transcription in eukaryotic cells. The nuclear receptor corepressor (N-CoR) and silencing mediator of retinoid and thyroid hormone receptors (SMRT) play a role in diverse transcriptional repression pathways. N-CoR and SMRT each exist in large protein complexes, and are thought to mediate repression, at least in part, by their ability to recruit histone deacetylases to generate repressive chromatin. HDAC3 is the main HDAC that associates with N-CoR and SMRT complexes, and its presence is thought to be essential for repression. TBLR1 is a WD-40 repeat protein that also associates with N-CoR and SMRT complexes. Although TBLR1 is thought to be a core component of both the N-CoR and SMRT complexes, its role in these complexes remains to be elucidated.
Specifications
| TBLR1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| TBL1XR1 | |
| 8030499H02Rik; A630076E03Rik; AW539987; C21; C230089I12Rik; DC42; F-box-like/WD repeat protein TBL1XR1; F-box-like/WD repeat-containing protein TBL1XR1; FLJ12894; Ira1; MRD41; nuclear receptor co-repressor/HDAC3 complex subunit; nuclear receptor corepressor/HDAC3 complex subunit TBLR1; RGD1560053; TBL1-related protein 1; TBL1XR1; TBLR1; transducin (beta)-like 1 X-linked receptor 1; transducin (beta)-like 1X-linked receptor 1; transducin beta-like 1X-related protein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 365755, 79718, 81004 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4935 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BHJ5, Q9BZK7 | |
| TBL1XR1 | |
| A synthesized peptide derived from human TBLR1 (430-500AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction