missing translation for 'onlineSavingsMsg'
Learn More

TBLR1 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£149.00 - £366.00

Specifications

Antigen TBLR1
Dilution Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18649041
View Documents
Novus Biologicals
NBP2-94126-0.02ml
0.02 mL
£149.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18624352
View Documents
Novus Biologicals
NBP2-94126-0.1ml
0.1 mL
£366.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TBLR1 Polyclonal antibody specifically detects TBLR1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

TBLR1
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Wnt Signaling Pathway
PBS (pH 7.3), 50% glycerol
79718
IgG
Affinity purified
Western Blot 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200
Polyclonal
Purified
RUO
Human, Mouse
C21, DC42, F-box-like/WD repeat-containing protein TBL1XR1, FLJ12894, IRA1Transducin beta-like 1X-related protein 1, nuclear receptor co-repressor/HDAC3 complex subunit, Nuclear receptor corepressor/HDAC3 complex subunit TBLR1, TBLR1TBL1-related protein 1, transducin (beta)-like 1 X-linked receptor 1, transducin (beta)-like 1X-linked receptor 1
Recombinant fusion protein containing a sequence corresponding to amino acids 120-170 of human TBLR1 (NP_078941.2). QQGSAKNGENTANGEENGAHTIANNHTDMMEVDGDVEIPPNKAVVLRGHES
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.