missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565664
This item is not returnable.
View return policy
Description
Immunogen sequence: RIRKEKPQQHN FTHRLLAAAL KSHSGNISCM DFSSNGKYLA TCADDRTIRI WSTKDFLQRE HRSMRANVEL DHATLVRFSP DCRAFIVWLA NGDTLRVFKM TKRE Highest antigen sequence identity to the following orthologs - mouse 96%, rat 98%.
This gene encodes a member of the beta-transducin protein family. Most proteins of the beta-transducin family are involved in regulatory functions. This protein is possibly involved in some intracellular signaling pathway. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.
Specifications
| TBL2 | |
| Polyclonal | |
| Unconjugated | |
| TBL2 | |
| C76179; DKFZP43N024; MGC134739; TBL2; transducin (beta)-like 2; transducin beta like 2; transducin beta-like protein 2; transducin -like 2; UNQ563/PRO1125; WBSCR13; Williams-Beuren syndrome chromosomal region 13 protein; Williams-Beuren syndrome chromosome region 13; WS beta-transducin repeats protein; WS-betaTRP; WS-bTRP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26608 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y4P3 | |
| TBL2 | |
| Recombinant Human TBL2. Recombinant protein control fragment (Product #RP-106338). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction