missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-69369
This item is not returnable.
View return policy
Description
TBL2 Polyclonal specifically detects TBL2 in Human samples. It is validated for Western Blot.
Specifications
| TBL2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| DKFZp434N024, DKFZP43N024, MGC134739, transducin (beta)-like 2, transducin beta-like protein 2, WBSCR13WS-betaTRPWS beta-transducin repeats protein, Williams-Beuren syndrome chromosomal region 13 protein, Williams-Beuren syndrome chromosome region 13 | |
| Rabbit | |
| 49 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Equine: 86%; Mouse: 85%, Rabbit: 86%. | |
| Human, Mouse, Equine, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9Y4P3 | |
| TBL2 | |
| Synthetic peptides corresponding to TBL2(transducin (beta)-like 2) The peptide sequence was selected from the N terminal of TBL2. Peptide sequence RSGRPACQKANGFPPDKSSGSKKQKQYQRIRKEKPQQHNFTHRLLAAALK. | |
| Affinity purified | |
| RUO | |
| 26608 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction