missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TBK1, GST Tag Recombinant Protein
TBK1 serine/threonine kinase is a member of the IKK family and is involved in NF-kappa B activation and IRF3 signaling.
£442.00 - £6900.00
Specifications
Accession Number | AAH34950.1 |
---|---|
For Use With (Application) | Kinase Assay |
KinaseFamily | CK1 Kinase Family |
Protein Length | Full Length |
Species | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15627888
|
Invitrogen™
A31513 |
10μg |
£442.00
10µg |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15637888
|
Invitrogen™
A31514 |
100μg |
£1602.00
100µg |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17188705
|
Invitrogen™
A31520 |
1mg |
£6900.00
1mg |
Estimated Shipment: 01-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This purified recombinant full-length protein can be used in a number of different applications, but it has been specifically validated for use with both the LanthaScreen™ & Z'-LYTE™ assay technologies to identify novel kinase inhibitors. TBL1 is also available in our SelectScreen™ Kinase Profiling Service.
- Validated for use with the LanthaScreen & Z'-LYTE assay technology
- Available in multiple pack sizes for convenient use
- Also available in our SelectSceen Kinase Profiling Service
Specifications
AAH34950.1 | |
CK1 Kinase Family | |
Human | |
GST Tag |
Kinase Assay | |
Full Length | |
Recombinant |