missing translation for 'onlineSavingsMsg'
Learn More

TBK1 Rabbit anti-Human, Mouse, Rat, Clone: 4G6C7, Novus Biologicals™

Product Code. 18336396 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336396 100 μg 100µL
18397104 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18336396 Supplier Novus Biologicals Supplier No. NBP316189100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TBK1 Monoclonal antibody specifically detects TBK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen TBK1
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone 4G6C7
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 2.7.11, EC 2.7.11.1, FLJ11330, NAK serine/threonine-protein kinase TBK 1, NAKserine/threonine-protein kinase TBK1, NF-kappa-B-activating kinase, T2K, TANK-binding kinase 1NF-kB-activating kinase, TBK 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TBK1 (Q9UHD2).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 29110
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.