Learn More
TBK1 Mouse anti-Human, Unconjugated, Clone: 2D7B1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67211-1-IG-20UL
This item is not returnable.
View return policy
Description
The TBK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TBK1. This antibody recognizes human, mouse, rat antigen. The TBK1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TBK1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunofluorescence, ELISA | |
| 2D7B1 | |
| Unconjugated | |
| 20 μL | |
| 29110 | |
| IgG2b |